Mouse anti-Golgi Complex, Clone: SPM581, Azide Free, Novus Biologicals™
Manufacturer:  Novus Biologicals Canada NBP23481301MG
 View more versions of this product
Catalog No. NBP23481301
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
Golgi Complex
SPM581
Unlabeled
10mM PBS. Does not contain BSA. Azide Free.
SU-DHL-1 large cell lymphoma cells.
Purified
RUO
Primary
Antibody recognizes Golgi complex in human cells.
Human
Immunocytochemistry,Immunofluorescence
1mg/mL
Purified
Murine
IgG1, κ
0.1mg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Protein A purified