Please login to your online account to display your discounted pricing

Mouse anti-Golgi Complex, Clone: 371-4, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453402MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453402

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Cross Reactivity Human
Antigen Golgi Complex
Isotype IgG1, κ
Quantity 0.2mg
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen SU-DHL-1 large cell lymphoma cells.
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Reconstitution Protein A purified
Clone 371-4
Concentration 1mg/mL
Conjugate Unlabeled