Please login to your online account to display your discounted pricing

Mouse anti-Golgi Complex, Clone: 371-4, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23453401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23453401

  • / Each
 

Description & Specifications

Specifications

Antigen Golgi Complex
Applications Immunocytochemistry
Applications Immunofluorescence
Clone 371-4
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen SU-DHL-1 large cell lymphoma cells.
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Target Species Human