Please login to your online account to display your discounted pricing

Mouse anti-GM-CSF R alpha, Clone: 4H1, PE, Novus Biologicals™

$197.00 - $362.00

Specifications

Antigen GM-CSF R alpha
Clone 4H1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD116.
Gene ID (Entrez) 1438
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP143823

 
novus biologicals canada
NBP143823100TESTS
100 Tests Each for $362.00

NBP14382325

 
novus biologicals canada
NBP14382325TESTS
25 Tests Each for $197.00
Description & Specifications

Specifications

Antigen GM-CSF R alpha
Clone 4H1
Concentration 0.2mg/mL
Immunogen The immunogen for this antibody was CD116.
Gene ID (Entrez) 1438
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1, κ
Conjugate PE
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry