Please login to your online account to display your discounted pricing

Mouse anti-Glypican 4, Clone: 4A3, Novus Biologicals™

$268.00 - $332.00

Specifications

Antigen Glypican 4
Clone 4A3
Concentration 1.0mg/mL
Immunogen Recombinant human GPC4 (401-529aa) purified from E. coli (NP_001439)
Gene Accession No. O75487
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14528601

 
novus biologicals canada
NBP14528601ML
0.1mL Each for $332.00

NBP14528605

 
novus biologicals canada
NBP145286005ML
0.05mL Each for $268.00
Description & Specifications

Specifications

Antigen Glypican 4
Clone 4A3
Concentration 1.0mg/mL
Immunogen Recombinant human GPC4 (401-529aa) purified from E. coli (NP_001439)
Gene Accession No. O75487
Gene ID (Entrez) 2239
Formulation PBS (pH 7.4). Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting