Please login to your online account to display your discounted pricing

Mouse anti-Glypican 4, Clone: 4A3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP145286005ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14528605

  • / Each
 

Description & Specifications

Specifications

Antigen Glypican 4
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Clone 4A3
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS (pH 7.4). Protein G purified.
Gene Accession No. O75487
Host Species Mouse
Immunogen Recombinant human GPC4 (401-529aa) purified from E. coli (NP_001439)
Isotype IgG1, κ
Purification Method Purified
Quantity 0.05mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2239
Test Specificity This antibody is specific to Human Glypican 4