Please login to your online account to display your discounted pricing

Mouse anti-Glypican 4, Clone: 4A3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14528601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14528601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Purified
Formulation PBS (pH 7.4). Protein G purified.
Immunogen Recombinant human GPC4 (401-529aa) purified from E. coli (NP_001439)
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2239
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Antigen Glypican 4
Gene Accession No. O75487
Host Species Mouse
Isotype IgG1, κ
Quantity 0.1mL
Test Specificity This antibody is specific to Human Glypican 4
Clone 4A3