Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: SPM183, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Glycophorin A
Clone SPM183
Immunogen Human erythrocytes treated with neuraminidase
Gene Accession No. P02724
Gene ID (Entrez) 2993
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23445001

 
novus biologicals canada
NBP23445001ML
0.1mL Each for $139.00

NBP23445005

 
novus biologicals canada
NBP23445005ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Glycophorin A
Clone SPM183
Immunogen Human erythrocytes treated with neuraminidase
Gene Accession No. P02724
Gene ID (Entrez) 2993
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Human
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)