Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: SPM183, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23445001ML

 View all options for this product


For Research Use Only

Catalog No. NBP23445001


Description & Specifications


Antigen Glycophorin A
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone SPM183
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P02724
Host Species Mouse
Immunogen Human erythrocytes treated with neuraminidase
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2993
Test Specificity Recognizes a sialoglycoprotein of 39kDa, identified as glycophorin A. It reacts with a peptide epitope on the extracellular domain of human glycophorin.