Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: HIR2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1431080025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14310803

  • / Each
 

Description & Specifications

Specifications

Antigen Glycophorin A
Applications Flow Cytometry
Clone HIR2
Concentration 0.5mg/mL
Conjugate Unlabeled
Format Affinity Purified
Formulation PBS, pH 7.2
Gene Accession No. P02724
Host Species Murine
Immunogen The immunogen for this antibody was CD235a.
Isotype IgG2b, κ
Purification Method Affinity Purified
Quantity 0.025mg
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2993
Target Species Human