Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: HIR2, Biotin, Novus Biologicals™

$175.00 - $329.00

Specifications

Antigen Glycophorin A
Clone HIR2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD235a.
Gene Accession No. P02724
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14346301

 
novus biologicals canada
NBP14346301MG
0.1mg Each for $329.00

NBP14346303

 
novus biologicals canada
NBP1434630025MG
0.025mg Each for $175.00
Description & Specifications

Specifications

Antigen Glycophorin A
Clone HIR2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD235a.
Gene Accession No. P02724
Gene ID (Entrez) 2993
Formulation Phosphate buffer pH 7.2
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Biotin
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry