Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: GPHN02, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446205

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Antigen Glycophorin A
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P02724
Host Species Mouse
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2993
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Immunogen Human erythrocytes treated with neuraminidase
Isotype IgM, κ
Purification Method Concentrated
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone GPHN02