Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: GPHN02, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446201ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446201

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Quantity 0.1mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Mouse
Antigen Glycophorin A
Regulatory Status RUO
Purification Method Concentrated
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Immunogen Human erythrocytes treated with neuraminidase
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Isotype IgM, κ
Gene Accession No. P02724
Clone GPHN02
Format Concentrated
Gene ID (Entrez) 2993