Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: A63-C/A9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446305ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446305

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Human
Host Species Mouse
Immunogen Human erythrocytes treated with neuraminidase
Purification Method Concentrated
Monoclonal or Polyclonal Monoclonal
Antigen Glycophorin A
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P02724
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Isotype IgM, κ
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 2993
Clone A63-C/A9
Conjugate Unlabeled
Regulatory Status RUO