Please login to your online account to display your discounted pricing

Mouse anti-Glycophorin A, Clone: A63-C/A9, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23446301ML

 View all options for this product


For Research Use Only

Catalog No. NBP23446301


Description & Specifications


Antigen Glycophorin A
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone A63-C/A9
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Gene Accession No. P02724
Host Species Mouse
Immunogen Human erythrocytes treated with neuraminidase
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2993