Please login to your online account to display your discounted pricing

Mouse anti-Glut1, Clone: SPM498, Novus Biologicals™

$307.00 - $551.93

Specifications

Antigen Glut1
Clone SPM498
Immunogen Synthetic peptide derived from C-terminus of human GLUT-1 protein.
Gene Accession No. P11166
Gene ID (Entrez) 6513
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13592605

 
novus biologicals canada
NBP13592605ML
0.5mL Each for $367.12

NBP13592601

 
novus biologicals canada
NBP13592601ML
0.1mL Each for $307.00

NBP13592610

 
novus biologicals canada
NBP13592610ML
1mL Each for $551.93
Description & Specifications

Specifications

Antigen Glut1
Clone SPM498
Immunogen Synthetic peptide derived from C-terminus of human GLUT-1 protein.
Gene Accession No. P11166
Gene ID (Entrez) 6513
Formulation PBS/1% BSA buffer pH 7.6 . Immunogen affinity purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2a, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)