Please login to your online account to display your discounted pricing

Mouse anti-Glut1, Clone: SPM498, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13592601ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13592601

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Primary or Secondary Primary
Regulatory Status RUO
Isotype IgG2a, κ
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Storage Requirements Store at 4°C. Do not freeze.
Quantity 0.1mL
Host Species Mouse
Antigen Glut1
Immunogen Synthetic peptide derived from C-terminus of human GLUT-1 protein.
Purification Method Affinity Purified
Clone SPM498
Conjugate Unlabeled
Formulation PBS/1% BSA buffer pH 7.6 . Immunogen affinity purified.
Gene Accession No. P11166
Test Specificity This product is specific for GLUT-1
Gene ID (Entrez) 6513