Please login to your online account to display your discounted pricing

Mouse anti-Ghrelin/Obestatin, Clone: 121, Novus Biologicals™

$406.15 - $406.15

Specifications

Antigen Ghrelin/Obestatin
Clone 121
Concentration 1.0mg/mL
Immunogen Synthetic ghrelin (14-28)
Gene Accession No. Q9UBU3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14718201

 
novus biologicals canada
NBP14718201MG
0.1mg Each for $406.15
Description & Specifications

Specifications

Antigen Ghrelin/Obestatin
Clone 121
Concentration 1.0mg/mL
Immunogen Synthetic ghrelin (14-28)
Gene Accession No. Q9UBU3
Gene ID (Entrez) 51738
Formulation 0.01M phosphate buffer, pH 7.4, containing 0.5M NaCl
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C in the dark.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgG2, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Western Blotting