Please login to your online account to display your discounted pricing

Mouse anti-Ghrelin/Obestatin, Clone: 121, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14718210MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14718210

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Western Blotting
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Gene Accession No. Q9UBU3
Host Species Mouse
Immunogen Synthetic ghrelin (14-28)
Quantity 1mg
Primary or Secondary Primary
Clone 121
Conjugate Unlabeled
Antigen Ghrelin/Obestatin
Formulation 0.01M phosphate buffer, pH 7.4, containing 0.5M NaCl
Isotype IgG2, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 51738
Test Specificity It binds human ghrelin in solution. It binds human ghrelin and cross-reacts with rat ghrelin. It is presumed to cross-react with ghrelins of other species with identical 14-28 sequences, such as mouse, dog and cat.
Reconstitution Protein A purified
Concentration 1.0mg/mL