Please login to your online account to display your discounted pricing

Mouse anti-Ghrelin/Obestatin, Clone: 121, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP14718201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14718201

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Test Specificity It binds human ghrelin in solution. It binds human ghrelin and cross-reacts with rat ghrelin. It is presumed to cross-react with ghrelins of other species with identical 14-28 sequences, such as mouse, dog and cat.
Concentration 1.0mg/mL
Formulation 0.01M phosphate buffer, pH 7.4, containing 0.5M NaCl
Quantity 0.1mg
Cross Reactivity Canine
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Antigen Ghrelin/Obestatin
Format Purified
Gene Accession No. Q9UBU3
Host Species Mouse
Applications ELISA
Applications Western Blotting
Immunogen Synthetic ghrelin (14-28)
Isotype IgG2, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C in the dark.
Primary or Secondary Primary
Gene ID (Entrez) 51738
Reconstitution Protein A purified
Clone 121
Conjugate Unlabeled