Please login to your online account to display your discounted pricing

Mouse anti-GFAP, Clone: SPM507, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen GFAP
Clone SPM507
Concentration 0.2mg/mL
Immunogen GFAP isolated from pig spinal cord
Gene Accession No. P14136
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23436501

 
novus biologicals canada
NBP23436501ML
0.1mL Each for $139.00

NBP23436505

 
novus biologicals canada
NBP23436505ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen GFAP
Clone SPM507
Concentration 0.2mg/mL
Immunogen GFAP isolated from pig spinal cord
Gene Accession No. P14136
Gene ID (Entrez) 2670
Formulation PBS with 0.05% BSA. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting