Please login to your online account to display your discounted pricing

Mouse anti-GFAP, Clone: SPM507, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23441302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23441302

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen GFAP isolated from pig spinal cord
Conjugate Unlabeled
Quantity 0.2mg
Test Specificity This MAb recognizes a protein of ∽50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP).
Host Species Mouse
Regulatory Status RUO
Primary or Secondary Primary
Clone SPM507
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 2670
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Isotype IgG1
Reconstitution Protein A purified
Monoclonal or Polyclonal Monoclonal
Purification Method Purified
Antigen GFAP
Gene Accession No. P14136