Please login to your online account to display your discounted pricing

Mouse anti-GFAP, Clone: SPM248, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23435305ML

 View all options for this product


For Research Use Only

Catalog No. NBP23435305


Description & Specifications


Antigen GFAP
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone SPM248
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA. Protein G purified.
Gene Accession No. P14136
Host Species Mouse
Immunogen GFAP isolated from pig spinal cord
Isotype IgG1
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2670
Test Specificity This MAb recognizes a protein of ∽50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins.