Please login to your online account to display your discounted pricing

Mouse anti-GFAP, Clone: SPM248, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Antigen GFAP
Formulation PBS with 0.05% BSA. Protein G purified.
Format Purified
Gene Accession No. P14136
Host Species Mouse
Immunogen GFAP isolated from pig spinal cord
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2670
Test Specificity This MAb recognizes a protein of ∽50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins.
Clone SPM248
Concentration 0.2mg/mL
Conjugate Unlabeled
Isotype IgG1