Please login to your online account to display your discounted pricing

Mouse anti-GFAP, Clone: SPM248, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440101MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440101

  • / Each
 

Description & Specifications

Specifications

Antigen GFAP
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Bovine
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rabbit
Cross Reactivity Rat
Gene Accession No. P14136
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen GFAP isolated from pig spinal cord
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This MAb recognizes a protein of ∽50kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins.
Concentration 1mg/mL
Conjugate Unlabeled
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Isotype IgG1
Primary or Secondary Primary
Gene ID (Entrez) 2670
Reconstitution Protein A purified
Clone SPM248
Format Purified