Please login to your online account to display your discounted pricing

Mouse anti-FUT3/Blood Group Lewis A, Clone: SPM522, Novus Biologicals™

$307.00 - $518.93

Specifications

Antigen FUT3/Blood Group Lewis A
Clone SPM522
Immunogen Mucin isolated from an ovarian cyst
Gene ID (Entrez) 2525
Formulation 10mM PBS/1% BSA buffer pH 7.4
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP13606001

 
novus biologicals canada
NBP13606001ML
0.1mL Each for $307.00

NBP13606010

 
novus biologicals canada
NBP13606010ML
1mL Each for $518.93
Description & Specifications

Specifications

Antigen FUT3/Blood Group Lewis A
Clone SPM522
Immunogen Mucin isolated from an ovarian cyst
Gene ID (Entrez) 2525
Formulation 10mM PBS/1% BSA buffer pH 7.4
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)