Please login to your online account to display your discounted pricing

Mouse anti-FUT3/Blood Group Lewis A, Clone: SPM522, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP13606070ML

 View all options for this product


For Research Use Only

Catalog No. NBP13606070


Description & Specifications


Antigen FUT3/Blood Group Lewis A
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Clone SPM522
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 10mM PBS/1% BSA buffer pH 7.4
Host Species Mouse
Immunogen Mucin isolated from an ovarian cyst
Isotype IgG1
Purification Method Purified
Quantity 7mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2525
Test Specificity This product is specific for FUT3/Blood Group Lewis A
Reconstitution Protein A purified