Please login to your online account to display your discounted pricing

Mouse anti-FUT3/Blood Group Lewis A, Clone: SPM522, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13606010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13606010

  • / Each
 

Description & Specifications

Specifications

Antigen FUT3/Blood Group Lewis A
Format Purified
Formulation 10mM PBS/1% BSA buffer pH 7.4
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Mucin isolated from an ovarian cyst
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Test Specificity This product is specific for FUT3/Blood Group Lewis A
Conjugate Unlabeled
Isotype IgG1
Purification Method Purified
Quantity 1mL
Storage Requirements Store at 4°C. Do not freeze.
Primary or Secondary Primary
Gene ID (Entrez) 2525
Clone SPM522
Reconstitution Protein A purified
Cross Reactivity Human