Please login to your online account to display your discounted pricing

Mouse anti-FUT3/Blood Group Lewis A, Clone: SPM522, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP13606001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP13606001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Antigen FUT3/Blood Group Lewis A
Formulation 10mM PBS/1% BSA buffer pH 7.4
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen Mucin isolated from an ovarian cyst
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 2525
Test Specificity This product is specific for FUT3/Blood Group Lewis A
Clone SPM522
Cross Reactivity Human
Isotype IgG1
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Conjugate Unlabeled