Please login to your online account to display your discounted pricing

Mouse anti-FSH R, Clone: 6E8.2F5, Novus Biologicals™

$109.00 - $259.02

Specifications

Antigen FSH R
Clone 6E8.2F5
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945]
Gene Accession No. P23945
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236489

 
novus biologicals canada
NBP236489
0.1mg Each for $259.02

NBP236489SS

 
novus biologicals canada
NBP236489SS
0.025mg Each for $109.00
Description & Specifications

Specifications

Antigen FSH R
Clone 6E8.2F5
Concentration 1.0mg/mL
Immunogen A synthetic peptide made to the human FSH R protein sequence (between residues 1-200). [Swiss-Prot P23945]
Gene Accession No. P23945
Gene ID (Entrez) 2492
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting