Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: SPM579, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23438505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23438505

  • / Each
 

Description & Specifications

Specifications

Antigen FoxP3
Host Species Mouse
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q9BZS1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Full-length human FOXP3 protein
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity Recognizes a protein of 47-55kDa, which is identified as FOXP3. Its precise epitope is not known, but it has been mapped to the N-terminal portion of the protein.
Clone SPM579
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate