Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: SPM579, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23438501ML

 View all options for this product


For Research Use Only

Catalog No. NBP23438501


Description & Specifications


Antigen FoxP3
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone SPM579
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q9BZS1
Host Species Mouse
Immunogen Full-length human FOXP3 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 50943
Test Specificity Recognizes a protein of 47-55kDa, which is identified as FOXP3. Its precise epitope is not known, but it has been mapped to the N-terminal portion of the protein.
Reconstitution Protein A purified