missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: SPM579, Azide Free, Novus Biologicals™

$396.83 - $698.00

Specifications

Antigen FoxP3
Clone SPM579
Concentration 1mg/mL
Immunogen Full-length human FOXP3 protein
Gene Accession No. Q9BZS1
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23443201 Novus Biologicals Canada
NBP23443201MG
0.1mg Each for $396.83
NBP23443202 Novus Biologicals Canada
NBP23443202MG
0.2mg Each for $698.00
Specifications Description & Specifications

Specifications

FoxP3
1mg/mL
Q9BZS1
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM579
Full-length human FOXP3 protein
50943
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation