Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: SPM579, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23443202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23443202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Full-length human FOXP3 protein
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes a protein of 47-55kDa, which is identified as FOXP3. Its precise epitope is not known, but it has been mapped to the N-terminal portion of the protein.
Reconstitution Protein A purified
Clone SPM579
Antigen FoxP3
Gene Accession No. Q9BZS1
Isotype IgG1, κ
Purification Method Purified
Gene ID (Entrez) 50943
Concentration 1mg/mL
Conjugate Unlabeled