Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: FXP3/197, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen FoxP3
Clone FXP3/197
Concentration 0.2mg/mL
Immunogen Full-length human FOXP3 protein
Gene Accession No. Q9BZS1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23428201

 
novus biologicals canada
NBP23428201ML
0.1mL Each for $139.00

NBP23428205

 
novus biologicals canada
NBP23428205ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen FoxP3
Clone FXP3/197
Concentration 0.2mg/mL
Immunogen Full-length human FOXP3 protein
Gene Accession No. Q9BZS1
Gene ID (Entrez) 50943
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation