Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: FXP3/197, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23428205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23428205

  • / Each
 

Description & Specifications

Specifications

Antigen FoxP3
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone FXP3/197
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q9BZS1
Host Species Mouse
Immunogen Full-length human FOXP3 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 50943
Reconstitution Protein A purified