Please login to your online account to display your discounted pricing

Mouse anti-FoxP3, Clone: FXP3/197, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23462001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23462001

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Antigen FoxP3
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Full-length human FOXP3 protein
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 50943
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. Q9BZS1
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Clone FXP3/197
Concentration 1mg/mL