Please login to your online account to display your discounted pricing

Mouse anti-FKBP52/FKBP4, Clone: KN382/EC1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19746502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19746502

  • / Each
 

Description & Specifications

Specifications

Antigen FKBP52/FKBP4
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone KN382/EC1
Conjugate Unlabeled
Format Purified
Formulation Liquid. In PBS, pH 7.2 containing 50% glycerol. Protein G purified.
Host Species Murine
Immunogen Partially purified 9S rabbit uterine progesterone receptor.
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2288
Target Species Bovine
Target Species Guinea Pig
Target Species Hamster
Target Species Human
Target Species Mink
Target Species Murine
Target Species Primate
Target Species Rabbit