Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: TV-1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450401MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Isotype IgG1, κ
Concentration 1mg/mL
Clone TV-1
Reconstitution Protein A purified
Gene Accession No. P02751
Quantity 0.1mg
Primary or Secondary Primary
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Antigen Fibronectin/Anastellin
Immunogen T-cell lymphoma biopsy
Gene ID (Entrez) 2335