Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM539, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23284801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23284801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Primary or Secondary Primary
Clone SPM539
Antigen Fibronectin/Anastellin
Formulation PBS with 0.05% BSA.
Gene Accession No. P02751
Host Species Mouse
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335
Reconstitution Protein A purified
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human