missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM539, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Fibronectin/Anastellin
Clone SPM539
Concentration 1mg/mL
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Gene Accession No. P02751
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23478501 Novus Biologicals Canada
NBP23478501MG
0.1mg Each for $384.00
NBP23478502 Novus Biologicals Canada
NBP23478502MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Fibronectin/Anastellin
1mg/mL
P02751
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
SPM539
Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
2335
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot