Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM539, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23478502MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23478502

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Conjugate Unlabeled
Concentration 1mg/mL
Host Species Mouse
Cross Reactivity Human
Regulatory Status RUO
Isotype IgG1, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Antigen Fibronectin/Anastellin
Gene Accession No. P02751
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Quantity 0.2mg
Purification Method Purified
Clone SPM539
Format Purified