Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM246, Novus Biologicals™

$139.00 - $429.00

Specifications

Antigen Fibronectin/Anastellin
Clone SPM246
Concentration 0.2mg/mL
Immunogen T-cell lymphoma biopsy
Gene Accession No. P02751
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23285010

 
novus biologicals canada
NBP23285010ML
1.0mL Each for $429.00

NBP23285001

 
novus biologicals canada
NBP23285001ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen Fibronectin/Anastellin
Clone SPM246
Concentration 0.2mg/mL
Immunogen T-cell lymphoma biopsy
Gene Accession No. P02751
Gene ID (Entrez) 2335
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting