Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM246, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285010ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285010

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Fibronectin/Anastellin
Formulation PBS with 0.05% BSA.
Immunogen T-cell lymphoma biopsy
Monoclonal or Polyclonal Monoclonal
Quantity 1.0mL
Host Species Mouse
Gene Accession No. P02751
Conjugate Unlabeled
Format Purified
Regulatory Status RUO
Isotype IgG1, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 2335
Concentration 0.2mg/mL
Primary or Secondary Primary
Reconstitution Protein A purified
Clone SPM246