Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM246, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23285001ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23285001

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Antigen Fibronectin/Anastellin
Formulation PBS with 0.05% BSA.
Gene Accession No. P02751
Immunogen T-cell lymphoma biopsy
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335
Reconstitution Protein A purified
Clone SPM246
Conjugate Unlabeled
Concentration 0.2mg/mL