Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: SPM246, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23474902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23474902

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Format Purified
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335
Primary or Secondary Primary
Quantity 0.2mg
Test Specificity Fibronectin is a soluble dimeric glycoprotein of 440kDa, which is present in cells, extracellular matrix, and blood. This MAb reacts with the cellular as well as plasma form of fibronectin.
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P02751
Regulatory Status RUO
Immunogen T-cell lymphoma biopsy
Purification Method Purified
Concentration 1mg/mL
Antigen Fibronectin/Anastellin
Host Species Mouse
Conjugate Unlabeled
Reconstitution Protein A purified
Clone SPM246
Isotype IgG1, κ
Formulation 10mM PBS. Does not contain BSA. Azide Free.