Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: HFN7.1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463302

  • / Each
 

Description & Specifications

Specifications

Antigen Fibronectin/Anastellin
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone HFN7.1
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P02751
Host Species Murine
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2335
Reconstitution Protein A purified
Target Species Human