Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: HFN7.1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463301

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Primary or Secondary Primary
Quantity 0.1mg
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Conjugate Unlabeled
Gene ID (Entrez) 2335
Regulatory Status RUO
Immunogen Human fibronectin purified from serum by affinity chromatography on gelatin-sepharose
Antigen Fibronectin/Anastellin
Concentration 1mg/mL
Isotype IgG1, κ
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Purification Method Purified
Gene Accession No. P02751
Clone HFN7.1
Reconstitution Protein A purified
Format Purified