Please login to your online account to display your discounted pricing

Mouse anti-Fibronectin/Anastellin, Clone: 2755-8, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23452601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23452601

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Antigen Fibronectin/Anastellin
Immunogen T-cell lymphoma biopsy
Gene Accession No. P02751
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Primary or Secondary Primary
Gene ID (Entrez) 2335
Concentration 1mg/mL
Isotype IgG1, κ
Purification Method Purified
Reconstitution Protein A purified
Clone 2755-8