Please login to your online account to display your discounted pricing

Mouse anti-FGF-23, Clone: FGF23/638, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23461701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23461701

  • / Each
 

Description & Specifications

Specifications

Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Clone FGF23/638
Cross Reactivity Human
Antigen FGF-23
Applications ELISA
Applications Functional Assay
Primary or Secondary Primary
Format Purified
Gene Accession No. Q9GZV9
Immunogen Recombinant human FGF23 protein
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Regulatory Status RUO
Gene ID (Entrez) 8074
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled